(7-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(7-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
OR111431-1G
OR111431-250MG
APOSOR111431-1GEA
186
GBP
APOSOR111431-1G
APOSOR111431-250MG
(7-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
(7-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
Formula:
C₉H₇BrN₂O₂ MW: 255,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11840279 CAS Number: 1217061-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...