1-(2,4-Dimethylphenyl)ethanol
Supplier: Apollo Scientific
|
Warning
|
OR111124-5G
OR111124-1G
APOSOR111124-5GEA
134
GBP
APOSOR111124-5G
APOSOR111124-1G
1-(2,4-Dimethylphenyl)ethanol
1-(2,4-Dimethylphenyl)ethanol
Formula:
C₁₀H₁₄O MW: 150,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00060890 CAS Number: 5379-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...