Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate

Supplier: Apollo Scientific
Warning

OR110756-1G OR110756-250MG
APOSOR110756-1GEA 142 GBP
APOSOR110756-1G APOSOR110756-250MG
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Formula: C₁₀H₉NO₄
Melting Pt: 247…253 °C
Storage Temperature: Ambient
MDL Number: MFCD07774184
CAS Number: 142166-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR