4-Chloroquinoline 97%

Supplier: Apollo Scientific
Warning

OR1107-1G OR1107-5G
APOSOR1107-1GEA 46 GBP
APOSOR1107-1G APOSOR1107-5G
4-Chloroquinoline 97%
4-Chloroquinoline
Formula: C₉H₆ClN
MW: 163,61 g/mol
Boiling Pt: 260…261 °C
Melting Pt: 28…31 °C
Density: 1,25 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006773
CAS Number: 611-35-8
EINECS: 210-267-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR