4-Hydroxy-3-nitrobenzonitrile

Supplier: Apollo Scientific

OR110136-10G OR110136-1G OR110136-5G
APOSOR110136-10GEA 127 GBP
APOSOR110136-10G APOSOR110136-1G APOSOR110136-5G
4-Hydroxy-3-nitrobenzonitrile
4-Hydroxy-3-nitrobenzonitrile
Formula: C₇H₄N₂O₃
MW: 164,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070775
CAS Number: 3272-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR