2,3,4-Trihydroxybenzoic acid

Supplier: Apollo Scientific
Warning

OR110134-1G OR110134-5G
APOSOR110134-1GEA 52.5 GBP
APOSOR110134-1G APOSOR110134-5G
2,3,4-Trihydroxybenzoic acid
2,3,4-Trihydroxybenzoic acid
Formula: C₇H₆O₅
MW: 170,12 g/mol
Melting Pt: 227…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00002447
CAS Number: 610-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR