3-Hydroxyphenylboronic acid pinacol ester 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

OR11013-5G OR11013-25G
APOSOR11013-5GEA 57.7 GBP
APOSOR11013-5G APOSOR11013-25G
3-Hydroxyphenylboronic acid pinacol ester 98%
3-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220,08 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD02093755
CAS Number: 214360-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR