3-Nitrobenzaldehyde

Supplier: Apollo Scientific
Warning

OR10999-250G OR10999-100G
APOSOR10999-250GEA 32.8 GBP
APOSOR10999-250G APOSOR10999-100G
3-Nitrobenzaldehyde
3-Nitrobenzaldehyde
Formula: C₇H₅NO₃
MW: 151.12 g/mol
Boiling Pt: 285…290 °C
Melting Pt: 55…59 °C
Density: 1.28 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007249
CAS Number: 99-61-6
EINECS: 202-772-6
Merck Index: 12,06684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR