(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinone

Supplier: Apollo Scientific
Warning

Synonyms: (4S,5R)-4-Methyl-5-phenyloxazolidin-2-one, (4S,5R)-(-)-4-Methyl-5-phenyloxazolidin-2-one

OR10970-1G OR10970-5G OR10970-250MG
APOSOR10970-1GEA 52.5 GBP
APOSOR10970-1G APOSOR10970-5G APOSOR10970-250MG
(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinone
(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinone
Formula: C₁₀H₁₁NO₂
MW: 177,2000 g/mol
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00066226
CAS Number: 16251-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR