N-(Hydroxymethyl)phthalimide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Hydroxymethyl)isoindoline-1,3-dione
OR10883-100G
OR10883-25G
OR10883-500G
APOSOR10883-100GEA
48.6
GBP
APOSOR10883-100G
APOSOR10883-25G
APOSOR10883-500G
N-(Hydroxymethyl)phthalimide
N-(Hydroxymethyl)phthalimide
Formula:
C₉H₇NO₃ MW: 177,16 g/mol Melting Pt: 147…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005899 CAS Number: 118-29-6 EINECS: 204-241-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...