4-(Hydroxymethyl)indole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(1H-Indol-4-yl)methanol
OR10506-1G
OR10506-250MG
APOSOR10506-1GEA
43.4
GBP
APOSOR10506-1G
APOSOR10506-250MG
4-(Hydroxymethyl)indole
4-(Hydroxymethyl)indole
Formula:
C₉H₉NO MW: 147,1739 g/mol Melting Pt: 46…50 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01632220 CAS Number: 1074-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...