4-(Dimethylcarbamoyl)phenylboronic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-(Dimethylcarbamoyl)phenyl)boronic acid, 4-(Dimethylcarbamoyl)benzeneboronic acid, 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid
OR10459-1G
OR10459-25G
OR10459-5G
APOSOR10459-1GEA
19.7
GBP
APOSOR10459-1G
APOSOR10459-25G
APOSOR10459-5G
4-(Dimethylcarbamoyl)phenylboronic acid 97%
4-(Dimethylcarbamoyl)phenylboronic acid
Formula:
C₉H₁₂BNO₃ MW: 193.01 g/mol Melting Pt: 130…136 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02258943 CAS Number: 405520-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...