2,3-Dihydrobenzofuran-5-boronic acid
Supplier: Apollo Scientific
|
Warning
|
OR10449-1G
OR10449-5G
APOSOR10449-1GEA
59.1
GBP
APOSOR10449-1G
APOSOR10449-5G
2,3-Dihydrobenzofuran-5-boronic acid
2,3-Dihydrobenzofuran-5-boronic acid
Formula:
C₈H₉BO₃ MW: 163,97 g/mol Melting Pt: 260…270 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02681979 CAS Number: 227305-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...