2-Chloro-5-nitrobenzonitrile
Supplier: Apollo Scientific
|
Warning
|
OR10416-100G
OR10416-25G
OR10416-5G
APOSOR10416-100GEA
33.6
GBP
APOSOR10416-100G
APOSOR10416-25G
APOSOR10416-5G
2-Chloro-5-nitrobenzonitrile
2-Chloro-5-nitrobenzonitrile
Formula:
C₇H₃ClN₂O₂ Melting Pt: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007292 CAS Number: 16588-02-6 EINECS: 240-643-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...