2-Bromophenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2-Bromophenyl)boronic acid, Dihydroxy(2-bromophenyl)borane, 2-Bromobenzeneboronic acid, (2-Bromophenyl)metaboric acid
OR10387-100G
OR10387-25G
APOSOR10387-100GEA
140
GBP
APOSOR10387-100G
APOSOR10387-25G
2-Bromophenylboronic acid 98%
2-Bromophenylboronic acid
Formula:
C₆H₆BBrO₂ MW: 200.83 g/mol Melting Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01114672 CAS Number: 244205-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...