Ethyl indole-2-carboxylate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester
OR10188-25G
OR10188-5G
APOSOR10188-25GEA
22.3
GBP
APOSOR10188-25G
APOSOR10188-5G
Ethyl indole-2-carboxylate 98%
Ethyl indole-2-carboxylate
Formula:
C₁₁H₁₁NO₂ MW: 189.21 g/mol Melting Pt: 122…125 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005609 CAS Number: 3770-50-1 EINECS: 223-206-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...