2-Thienylacetyl chloride 96%

Supplier: Apollo Scientific
Danger

Synonyms: 2-Thiopheneacetyl chloride, Thiophene-2-acetyl chloride

OR10151-100G OR10151-25G
APOSOR10151-100GEA 132 GBP
APOSOR10151-100G APOSOR10151-25G
2-Thienylacetyl chloride 96%
2-Thienylacetyl chloride
Formula: C₆H₅ClOS
MW: 160,62 g/mol
Boiling Pt: 130…135 °C/90mmHg
Density: 1,303 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005456
CAS Number: 39098-97-0
EINECS: 254-290-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR