5-Chloro-2-pentanone 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Chloropentan-2-one
OR10086-100G
OR10086-25G
OR10086-250G
APOSOR10086-100GEA
19.7
GBP
APOSOR10086-100G
APOSOR10086-25G
APOSOR10086-250G
5-Chloro-2-pentanone 97%
5-Chloro-2-pentanone
Formula:
C₅H₉ClO MW: 120,58 g/mol Boiling Pt: 63…64 °C/12mmHg Density: 1,057 g/cm³ (25 °C) Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001008 CAS Number: 5891-21-4 EINECS: 227-565-8 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...