1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N-(2-Chloroethyl)pyrrolidine hydrochloride
OR0992-25G
OR0992-5G
OR0992-10G
APOSOR0992-25GEA
48.6
GBP
APOSOR0992-25G
APOSOR0992-5G
APOSOR0992-10G
1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
1-(2-Chloroethyl)pyrrolidine hydrochloride
Formula:
C₆H₁₂ClN MW: 170,08 g/mol Melting Pt: 169…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012718 CAS Number: 7250-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...