Pyrrolidine 99%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Tetrahydropyrrole
APOSOR0971-100MLEA
15.8
GBP
APOSOR0971-100ML
Pyrrolidine 99%
Pyrrolidine
Formula:
C₄H₉N MW: 71,12 g/mol Boiling Pt: 86…88 °C Melting Pt: –60 °C Density: 0,856 g/cm³ (25 °C) Flash Pt: 3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005249 CAS Number: 123-75-1 EINECS: 204-648-7 UN: 1922 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...