4-Methyl-1,4'-bipiperidine hydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR0887-1G
OR0887-5G
APOSOR0887-1GEA
30.2
GBP
APOSOR0887-1G
APOSOR0887-5G
4-Methyl-1,4'-bipiperidine hydrochloride
4-Methyl-1,4'-bipiperidine hydrochloride
Formula:
C₁₁H₂₃ClN₂ MW: 218,77 g/mol Melting Pt: 40…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD17676095 CAS Number: 116797-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...