Methyl-3-amino-5-nitrobenzoate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 3-amino-5-nitrobenzoate
OR0861-10G
OR0861-2G
APOSOR0861-10GEA
492
GBP
APOSOR0861-10G
APOSOR0861-2G
Methyl-3-amino-5-nitrobenzoate
Methyl 3-amino-5-nitrobenzoate
Formula:
C₈H₈N₂O₄ MW: 196,1600 g/mol Melting Pt: 165…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00461253 CAS Number: 23218-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...