2-Bromo-1-indanol 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-1-hydroxyindane
OR0846-10G
OR0846-250G
OR0846-50G
APOSOR0846-10GEA
0
GBP
APOSOR0846-10G
APOSOR0846-250G
APOSOR0846-50G
2-Bromo-1-indanol 99%
2-Bromo-1-indanol
Formula:
C₉H₉BrO MW: 213,07 g/mol Melting Pt: 130…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003798 CAS Number: 5400-80-6 EINECS: 226-442-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...