Diphenyl disulphide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,2-Diphenyldisulphane, Phenyl disulphide
OR0792-100G
OR0792-25G
OR0792-500G
APOSOR0792-100GEA
106
GBP
APOSOR0792-100G
APOSOR0792-25G
APOSOR0792-500G
Diphenyl disulphide
Diphenyl disulphide
Formula:
C₁₂H₁₀S₂ MW: 218,34 g/mol Boiling Pt: 191…192 °C/15mmHg Melting Pt: 58…62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003065 CAS Number: 882-33-7 EINECS: 212-926-4 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...