(S)-(-)-Indoline-2-carboxylic acid
Supplier: Apollo Scientific
Synonyms:
(S)-Indoline-2-carboxylic acid
OR0737-100G
OR0737-25G
APOSOR0737-100GEA
335
GBP
APOSOR0737-100G
APOSOR0737-25G
(S)-(-)-Indoline-2-carboxylic acid
(S)-(-)-Indoline-2-carboxylic acid
Formula:
C₉H₉NO₂ MW: 163,18 g/mol Melting Pt: 178…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00070578 CAS Number: 79815-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...