(S)-2-Amino-3-cyclohexylpropanoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Cyclohexyl-L-alanine
OR0736-1G
OR0736-25G
OR0736-5G
APOSOR0736-1GEA
46
GBP
APOSOR0736-1G
APOSOR0736-25G
APOSOR0736-5G
(S)-2-Amino-3-cyclohexylpropanoic acid
(S)-2-Amino-3-cyclohexylpropanoic acid
Formula:
C₉H₁₇NO₂ MW: 171,24 g/mol Melting Pt: 322…328 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00153484 CAS Number: 27527-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...