(2-Bromoethoxy)benzene 98%

Supplier: Apollo Scientific
Warning

Synonyms: β-Bromophenetole, 2-Phenoxyethyl bromide, 2-Bromoethyl phenyl ether, ω-Bromophenetole

OR0698-25G OR0698-100G OR0698-500G
APOSOR0698-25GEA 13.2 GBP
APOSOR0698-25G APOSOR0698-100G APOSOR0698-500G
(2-Bromoethoxy)benzene 98%
(2-Bromoethoxy)benzene
Formula: BrCH₂CH₂OC₆H₅
MW: 201,06 g/mol
Boiling Pt: 135…137 °C/20mmHg
Melting Pt: 31…35 °C
Density: 1,35 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000234
CAS Number: 589-10-6
EINECS: 209-634-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR