3-Hydroxy-1-adamantane carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 3-Hydroxyadamantane-1-carboxylic acid, 3-Hydroxy-1-adamantanecarboxylic acid

OR0509-10G OR0509-25G OR0509-5G
APOSOR0509-10GEA 26.3 GBP
APOSOR0509-10G APOSOR0509-25G APOSOR0509-5G
3-Hydroxy-1-adamantane carboxylic acid
3-Hydroxy-1-adamantane carboxylic acid
Formula: C₁₁H₁₆O₃
MW: 196,25 g/mol
Melting Pt: 203…207 °C
Density: 1,377 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00193929
CAS Number: 42711-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR