(3,4-Dichlorophenyl)acetic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3,4-Dichlorophenylacetic acid

OR0359-100G OR0359-500G
APOSOR0359-100GEA 72.2 GBP
APOSOR0359-100G APOSOR0359-500G
(3,4-Dichlorophenyl)acetic acid 98%
(3,4-Dichlorophenyl)acetic acid
Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205,04 g/mol
Melting Pt: 87…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00004333
CAS Number: 5807-30-7
EINECS: 227-368-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR