2-Chloro-5-propylpyrimidine 97%
Supplier: Apollo Scientific
|
Warning
|
OR0295-10G
OR0295-1G
APOSOR0295-10GEA
94.4
GBP
APOSOR0295-10G
APOSOR0295-1G
2-Chloro-5-propylpyrimidine 97%
2-Chloro-5-propylpyrimidine
Formula:
C₇H₉ClN₂ Boiling Pt: 100 °C/3mmHg Density: 1,142 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00799403 CAS Number: 219555-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...