3-(4'-Chlorophenyl)propanol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(4-Chlorophenyl)propan-1-ol
OR0287-1G
OR0287-25G
OR0287-5G
APOSOR0287-1GEA
19.7
GBP
APOSOR0287-1G
APOSOR0287-25G
APOSOR0287-5G
3-(4'-Chlorophenyl)propanol 95%
3-(4'-Chlorophenyl)propanol
Formula:
C₉H₁₁ClO MW: 170,64 g/mol Boiling Pt: 98…102 °C/0,1mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00831102 CAS Number: 6282-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...