3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR0284-10G
OR0284-250G
OR0284-50G
APOSOR0284-10GEA
59.1
GBP
APOSOR0284-10G
APOSOR0284-250G
APOSOR0284-50G
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Formula:
C₁₁H₈ClNO₃ MW: 237,64 g/mol Melting Pt: 187…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020813 CAS Number: 23598-72-3 EINECS: 245-770-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...