5-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

OR01956-1G OR01956-250MG
APOSOR01956-1GEA 197 GBP
APOSOR01956-1G APOSOR01956-250MG
5-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
5-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₈H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09836168
CAS Number: 1000017-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR