4-Allyltoluene

Supplier: Apollo Scientific
Warning

Synonyms: p-Allyltoluene

OR01922-5G OR01922-1G
APOSOR01922-5GEA 128 GBP
APOSOR01922-5G APOSOR01922-1G
4-Allyltoluene
4-Allyltoluene
Formula: C₁₀H₁₂
MW: 132,21 g/mol
Boiling Pt: 86…87 °C/35mmHg
Density: 0,865 g/cm³ (25 °C)
Flash Pt: 54,4 °C
Storage Temperature: Ambient
CAS Number: 3333-13-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR