1H-Benzimidazole-5-carboxylic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1H-Benzo[d]imidazole-5-carboxylic acid, Benzimidazole-5-carboxylic acid, 5-Benzimidazolecarboxylic acid
OR01704-250G
OR01704-25G
OR01704-50G
APOSOR01704-250GEA
597
GBP
APOSOR01704-250G
APOSOR01704-25G
APOSOR01704-50G
1H-Benzimidazole-5-carboxylic acid 98%
1H-Benzimidazole-5-carboxylic acid
Formula:
C₈H₆N₂O₂ MW: 162,15 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011555 CAS Number: 15788-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...