2-Amino-4,6-dimethylnicotinamide
Supplier: Apollo Scientific
|
Warning
|
OR017-1G
OR017-500MG
APOSOR017-1GEA
410
GBP
APOSOR017-1G
APOSOR017-500MG
2-Amino-4,6-dimethylnicotinamide
2-Amino-4,6-dimethylnicotinamide
Formula:
C₈H₁₁N₃O Melting Pt: 172…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00138572 CAS Number: 7144-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...