4-[(4-Bromophenyl)sulphonyl]piperidine hydrochloride, Technical Grade
Supplier: Apollo Scientific
|
Warning
|
OR01646-1G
OR01646-5G
APOSOR01646-1GEA
328
GBP
APOSOR01646-1G
APOSOR01646-5G
4-[(4-Bromophenyl)sulphonyl]piperidine hydrochloride, Technical Grade
4-[(4-Bromophenyl)sulphonyl]piperidine hydrochloride
Formula:
C₁₁H₁₅BrClNO₂S MW: 340.67 g/mol Melting Pt: 279…283 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04116083 CAS Number: 226400-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...