Methyl-3-(aminocarbonyl)benzoate 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 3-carbamoylbenzoate
OR0163-1G
OR0163-250MG
OR0163-5G
APOSOR0163-1GEA
97.1
GBP
APOSOR0163-1G
APOSOR0163-250MG
APOSOR0163-5G
Methyl-3-(aminocarbonyl)benzoate 95+%
Methyl-3-(aminocarbonyl)benzoate
Formula:
C₉H₉NO₃ MW: 179,18 g/mol Melting Pt: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD06245500 CAS Number: 106748-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...