Imidazo[1,2-a]pyrimidine

Supplier: Apollo Scientific
Warning

OR01549-1G OR01549-5G
APOSOR01549-1GEA 68.3 GBP
APOSOR01549-1G APOSOR01549-5G
Imidazo[1,2-a]pyrimidine
Imidazo[1,2-a]pyrimidine
Formula: C₆H₅N₃
MW: 119,1200 g/mol
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD06245377
CAS Number: 274-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR