Cyromazine

Supplier: Apollo Scientific

Synonyms: 2,4-Diamino-6-(cyclopropylamino)-1,3,5-triazine, N2-Cyclopropylmelamine

OR015047-25G OR015047-5G
APOSOR015047-25GEA 52.5 GBP
APOSOR015047-25G APOSOR015047-5G
Cyromazine
Cyromazine
Formula: C₆H₁₀N₆
MW: 166,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078650
CAS Number: 66215-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR