4-Chlorophenyl cyanomethyl sulphone 98%
Supplier: Apollo Scientific
|
Warning
|
OR0123-5G
OR0123-1G
OR0123-10G
APOSOR0123-5GEA
112
GBP
APOSOR0123-5G
APOSOR0123-1G
APOSOR0123-10G
4-Chlorophenyl cyanomethyl sulphone 98%
4-Chlorophenyl cyanomethyl sulphone
Formula:
C₈H₆ClNO₂S Melting Pt: 168…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045626 CAS Number: 1851-09-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...