Boc-Arg(NO₂)-OH

Supplier: Apollo Scientific
Warning

Synonyms: Nα-Boc-Nomega-nitro-L-arginine

OR0122-100G OR0122-25G OR0122-500G
APOSOR0122-100GEA 206 GBP
APOSOR0122-100G APOSOR0122-25G APOSOR0122-500G
Boc-Arg(NO₂)-OH
Boc-Arg(NO₂)-OH
Formula: C₁₁H₂₁N₅O₆
MW: 319,318 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065556
CAS Number: 2188-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR