2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
PBD, 2-([1,1-Biphenyl]-4-yl)-5-(4-(tert-butyl)phenyl)-1,3,4-oxadiazole, 2-(4-Biphenylyl)-5-(4-tert-butylphenyl)-1,3,4-oxadiazole
OR0007-100G
OR0007-250G
APOSOR0007-100GEA
95.7
GBP
APOSOR0007-100G
APOSOR0007-250G
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole
2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole
Formula:
C₂₄H₂₂N₂O MW: 354,45 g/mol Melting Pt: 136…138 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003101 CAS Number: 15082-28-7 EINECS: 239-135-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...