Rubidium bromide 99,9%

Supplier: Apollo Scientific

IN3013-25G IN3013-5G IN3013-100G
APOSIN3013-25GEA 233 GBP
APOSIN3013-25G APOSIN3013-5G APOSIN3013-100G
Rubidium bromide 99,9%
Rubidium bromide
Formula: BrRb
Melting Pt: 693 °C
Storage Temperature: Ambient
MDL Number: MFCD00011186
CAS Number: 7789-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR