Nickel(II) acetylacetonate, anhydrous
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate
IN2707-25G
IN2707-250G
APOSIN2707-25GEA
125
GBP
APOSIN2707-25G
APOSIN2707-250G
Nickel(II) acetylacetonate, anhydrous
Nickel (II) acetylacetonate
Formula:
C₁₀H₁₄NiO₄ MW: 256,91 g/mol Melting Pt: 230 °C Density: 8,9 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000024 CAS Number: 3264-82-2 EINECS: 221-875-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...