Aluminium nitrate hydrate 99.999%
Supplier: Apollo Scientific
|
Warning
|
IN1027-100G
IN1027-500G
APOSIN1027-100GEA
165
GBP
APOSIN1027-100G
APOSIN1027-500G
Aluminium nitrate hydrate 99.999%
Aluminium nitrate hydrate
Formula:
AlN₃O₉·H₂O MW: 231,01 g/mol Boiling Pt: 130 °C Melting Pt: 73,5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003419 CAS Number: 25838-59-9 UN: 1438 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...