Zearalenone

Supplier: Apollo Scientific
Danger

Synonyms: Benzoxacyclotetradecin-1,7(8H)-dione, (S,E)-14,16-Dihydroxy-3-methyl-3,4,5,6,9,10-hexahydro-1H-benzo[c][1]oxacyclotetradecine-1,7(8H)-dione

BIZ0112-25MG BIZ0112-50MG BIZ0112-100MG
APOSBIZ0112-25MGEA 125 GBP
APOSBIZ0112-25MG APOSBIZ0112-50MG APOSBIZ0112-100MG
Zearalenone
Zearalenone

Fungal mycotoxin.

Formula: C₁₈H₂₂O₅
MW: 318,4 g/mol
Melting Pt: 161…162 °C
Storage Temperature: Freezer
CAS Number: 17924-92-4
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR