SU 3327
Supplier: Apollo Scientific
Synonyms:
5-((5-Nitrothiazol-2-yl)thio)-1,3,4-thiadiazol-2-amine, SU3327
BISN0309-5MG
BISN0309-10MG
BISN0309-25MG
APOSBISN0309-5MGEA
145
GBP
APOSBISN0309-5MG
APOSBISN0309-10MG
APOSBISN0309-25MG
SU 3327
SU 3327
MAPK/ERK Signalling Pathway
Formula:
C₅H₃N₅O₂S₃ MW: 261,30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01927019 CAS Number: 40045-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...