Refametinib
Supplier: Apollo Scientific
Synonyms:
BAY 86-9766, (S)-N-(3,4-Difluoro-2-((2-fluoro-4-iodophenyl)amino)-6-methoxyphenyl)-1-(2,3-dihydroxypropyl)cyclopropane-1-sulphonamide, N-[3,4-Difluoro-2-(2-fluoro-4-iodoanilino)-6-methoxyphenyl]-1-[(2S)-2,3-dihydroxypropyl]cyclopropane-1-sulphonamide
BISN0230-1MG
BISN0230-5MG
BISN0230-10MG
APOSBISN0230-1MGEA
78.7
GBP
APOSBISN0230-1MG
APOSBISN0230-5MG
APOSBISN0230-10MG
Refametinib
Refametinib
MAPK/ERK Signalling Pathway
Formula:
C₁₉H₂₀F₃IN₂O₅S MW: 572,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18633256 CAS Number: 923032-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...