Sorafenib, p-toluenesulphonate salt, [3H]
Supplier: Apollo Scientific
Synonyms:
Sorafenib tosylate [3H]
BISN0216-10MG
BISN0216-50MG
BISN0216-100MG
APOSBISN0216-10MGEA
59.1
GBP
APOSBISN0216-10MG
APOSBISN0216-50MG
APOSBISN0216-100MG
Sorafenib, p-toluenesulphonate salt, [3H]
Sorafenib, p-toluenesulphonate salt, [3H]
MAPK/ERK Signalling Pathway
Storage Temperature:
Ambient |
CAS Number:
475207-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...